Assay Details
| Gene Symbol: Mlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF449425.1 | - | 1 - 2 | 498 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF449425.1 transcripts.
Set Membership: |
|

Mlc1
Genomic Map