Assay Details
| Gene Symbol: Ntng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB052336.1 | - | 3 - 4 | 1030 | 86 | |
| AF475079.1 | - | 3 - 4 | 1030 | 86 | ||
| BC141258.1 | - | 3 - 4 | 1089 | 9055870 | 86 | |
| BC145600.1 | - | 3 - 4 | 1089 | 9054166 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ntng2
Genomic Map