Assay Details
| Gene Symbol: Tubgcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286007.1 | NP_001272936.1 | 13 - 14 | 2383 | 60 | |
| NM_001286009.1 | NP_001272938.1 | 13 - 14 | 2383 | 60 | ||
| NM_001286011.1 | NP_001272940.1 | 12 - 13 | 2150 | 60 | ||
| NM_133755.2 | NP_598516.2 | 13 - 14 | 2310 | 60 | ||
| GenBank mRNA | AK006233.1 | - | 3 - 4 | 414 | 60 | |
| AK045084.1 | - | 13 - 14 | 2099 | 60 | ||
| AK146771.1 | - | 13 - 14 | 2305 | 60 | ||
| AK158060.1 | - | 13 - 14 | 2099 | 60 | ||
| AK167924.1 | - | 13 - 14 | 2205 | 60 | ||
| BC012519.1 | - | 13 - 14 | 2238 | 3601589 | 60 | |
| BC025564.1 | - | 12 - 13 | 2072 | 60 | ||
| BC025582.1 | - | 13 - 14 | 2199 | 5324981 | 60 | |
| BC027034.1 | - | 12 - 13 | 2584 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp2
Genomic Map