Assay Details
| Gene Symbol: Ptov1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133949.1 | NP_598710.1 | 4 - 5 | 719 | 76 | |
| GenBank mRNA | AK184608.1 | - | 3 - 4 | 76 | ||
| AK211289.1 | - | 3 - 4 | 194 | 76 | ||
| AK211387.1 | - | 3 - 4 | 76 | |||
| BC008543.1 | - | 4 - 5 | 595 | 3496037 | 76 | |
| BC024632.1 | - | 4 - 5 | 719 | 4222241 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptov1
Genomic Map