Assay Details
| Gene Symbol: Vmn1r237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134200.1 | NP_598961.1 | 1 | 836 | 81 | |
| GenBank mRNA | BC125543.1 | - | 1 | 890 | 40130058 | 81 |
| BC138066.1 | - | 1 | 890 | 8861085 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082549 | NM_134200.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082549 | BC125543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082549 | BC138066.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737849 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166869 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082549 | NM_134200.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082549 | BC125543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082549 | BC138066.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P283384 | NM_134200.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P283384 | BC125543.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P283384 | BC138066.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31085774 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166869 | - | - |
| Whole Mouse Genome G4122A | A_51_P283384 | NM_134200.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P283384 | BC125543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P283384 | BC138066.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116491 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13201444 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13201443 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13201425 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13201442 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13201429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13201428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31085753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13201425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13589900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13201427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13589896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13239573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31085756 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442193 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442191 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 792328 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3060136 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3060136 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003060136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2507665 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2507665 | - | - |
| mouse-6 v1.1 (Illumina) | scl8628.1.1_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3060136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl8628.1.1_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2507665 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2507665 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl8628.1.1_3-S | - | - |

Vmn1r237
Genomic Map