Assay Details
| Gene Symbol: Casc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ292072.1 | - | 85 | 64 | ||
| Gene Symbol: Gm12359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087290.1 | - | 1 - 2 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ292072.1 transcripts.
Set Membership: |
|

Casc3
Genomic Map