Assay Details
| Gene Symbol: Sardh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138665.2 | NP_619606.1 | 14 - 15 | 2487 | 66 | |
| GenBank mRNA | AK140451.1 | - | 13 - 14 | 1764 | 66 | |
| AK159236.1 | - | 14 - 15 | 2451 | 66 | ||
| AK159681.1 | - | 14 - 15 | 2451 | 66 | ||
| AK159704.1 | - | 14 - 15 | 2451 | 66 | ||
| AK163515.1 | - | 14 - 15 | 2016 | 66 | ||
| AK163660.1 | - | 13 - 14 | 1763 | 66 | ||
| AK178634.1 | - | 2 - 3 | 235 | 66 | ||
| AK183193.1 | - | 2 - 3 | 223 | 66 | ||
| BC003456.1 | - | 14 - 15 | 1996 | 2648344 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sardh
Genomic Map