Assay Details
| Gene Symbol: Rassf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138956.3 | NP_620406.1 | 2 - 3 | 352 | 72 | |
| GenBank mRNA | AF332864.1 | - | 2 - 3 | 363 | 72 | |
| AK155495.1 | - | 2 - 3 | 379 | 72 | ||
| AK167627.1 | - | 2 - 3 | 185 | 72 | ||
| BC011511.1 | - | 2 - 3 | 352 | 3495833 | 72 | |
| BC055023.1 | - | 2 - 3 | 407 | 5250629 | 72 | |
| BC071204.1 | - | 2 - 3 | 360 | 2648929 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rassf3
Genomic Map