Assay Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139140.1 | NP_631879.1 | 2 - 3 | 336 | 87 | |
| GenBank mRNA | AB079132.1 | - | 2 - 3 | 336 | 87 | |
| AK048720.1 | - | 2 - 3 | 378 | 87 | ||
| AK135838.1 | - | 2 - 3 | 394 | 87 | ||
| AK159335.1 | - | 2 - 3 | 430 | 87 | ||
| BC072579.1 | - | 2 - 3 | 603 | 87 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB079132.1,AK048720.1,AK135838.1,NM_139140.1 transcripts.
Set Membership: |
|

Spats2
Genomic Map