Assay Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139140.1 | NP_631879.1 | 5 - 6 | 581 | 85 | |
| GenBank mRNA | AB079132.1 | - | 5 - 6 | 581 | 85 | |
| AK048720.1 | - | 5 - 6 | 623 | 85 | ||
| AK135838.1 | - | 5 - 6 | 639 | 85 | ||
| AK159335.1 | - | 5 - 6 | 86 | |||
| AK167788.1 | - | 4 - 5 | 86 | |||
| AK171266.1 | - | 4 - 5 | 86 | |||
| BC023432.1 | - | 3 - 4 | 231 | 5029821 | 85 | |
| BC072579.1 | - | 5 - 6 | 848 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spats2
Genomic Map