Assay Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139140.1 | NP_631879.1 | 6 - 7 | 839 | 69 | |
| GenBank mRNA | AB079132.1 | - | 6 - 7 | 839 | 69 | |
| AK048720.1 | - | 6 - 7 | 881 | 69 | ||
| AK135838.1 | - | 6 - 7 | 897 | 69 | ||
| AK159335.1 | - | 6 - 7 | 934 | 69 | ||
| AK167788.1 | - | 5 - 6 | 517 | 69 | ||
| AK171266.1 | - | 5 - 6 | 595 | 69 | ||
| BC023432.1 | - | 4 - 5 | 489 | 5029821 | 69 | |
| BC072579.1 | - | 6 - 7 | 1106 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spats2
Genomic Map