Assay Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289709.1 | NP_001276638.1 | 3 - 4 | 447 | 100 | |
| NM_001289710.1 | NP_001276639.1 | 3 - 4 | 447 | 100 | ||
| NM_139302.2 | NP_647463.1 | 3 - 4 | 447 | 100 | ||
| GenBank mRNA | AF426314.1 | - | 3 - 4 | 344 | 100 | |
| AK129455.1 | - | 3 - 4 | 375 | 100 | ||
| AK152409.1 | - | 3 - 4 | 377 | 100 | ||
| AK152502.1 | - | 3 - 4 | 398 | 100 | ||
| AK155248.1 | - | 3 - 4 | 372 | 100 | ||
| AK167762.1 | - | 3 - 4 | 420 | 100 | ||
| AK188040.1 | - | 2 - 3 | 157 | 100 | ||
| BC024477.1 | - | 3 - 4 | 362 | 4977001 | 100 | |
| BC028859.1 | - | 3 - 4 | 436 | 4167974 | 100 | |
| CX216029.1 | - | 3 - 4 | 437 | 100 | ||
| EF676083.1 | - | 3 - 4 | 423 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3glb2
Genomic Map