Assay Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289709.1 | NP_001276638.1 | 5 - 6 | 674 | 79 | |
| NM_139302.2 | NP_647463.1 | 5 - 6 | 674 | 79 | ||
| GenBank mRNA | AF426314.1 | - | 5 - 6 | 571 | 79 | |
| AK129455.1 | - | 5 - 6 | 602 | 79 | ||
| AK152409.1 | - | 5 - 6 | 604 | 79 | ||
| AK155248.1 | - | 5 - 6 | 599 | 79 | ||
| AK167762.1 | - | 5 - 6 | 647 | 79 | ||
| BC024477.1 | - | 5 - 6 | 589 | 4977001 | 79 | |
| EF676083.1 | - | 5 - 6 | 650 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102719 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102720 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971849 | AK129455.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971849 | BC024477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971849 | EF676083.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971849 | AK152502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971849 | AK155248.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152521 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971849 | AK129455.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971849 | BC024477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971849 | EF676083.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971849 | AK152502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971849 | AK155248.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297647 | AK129455.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297647 | BC024477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297647 | EF676083.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297647 | AK152502.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P297647 | AK155248.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P160209 | AK129455.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P160209 | BC024477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P160209 | BC028859.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P160209 | EF676083.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P160209 | AK152502.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21102701 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00334088 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00334062 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00334072 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04470745 | - | - |
| Whole Mouse Genome G4122A | A_51_P297647 | AK129455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297647 | BC024477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297647 | EF676083.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297647 | AK152502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297647 | AK155248.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160209 | AK129455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160209 | BC024477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160209 | BC028859.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160209 | EF676083.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160209 | AK152502.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101365 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00334072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01806866 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01841924 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01841912 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01830639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102720 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102719 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21102721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21102653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01813242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01713113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01623499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01623509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01197772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01623502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01649012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01939467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01623504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01939464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02179445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01649013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01623508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01931333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01496655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01713132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01939462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01939466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01623503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01623507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01702091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01713112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02107900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01980789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21102667 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10481466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481460 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481463 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481472 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481461 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481462 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481470 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481469 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481473 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481467 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481471 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426499_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10481457 | - | - |
| Mouse Genome 430 2.0 Array | 1426499_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426499_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 227700_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 227700_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200027638.mm.1 | NM_001289709.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027638.mm.1 | NM_139302.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200027638.mm.1 | TC0200003269.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426499_PM_at | - | - |
| Mouse Expression Set 430 | 1426499_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426499_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426499_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426499_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426499_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 921421 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2030086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2635240 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2635240 | - | - |
| mouse-6 v1.1 (Illumina) | scl19472.10_43-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19472.10_43-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030086 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2635240 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2635240 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl19472.10_43-S | - | - |

Sh3glb2
Genomic Map