Assay Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289709.1 | NP_001276638.1 | 6 - 7 | 738 | 85 | |
| NM_139302.2 | NP_647463.1 | 6 - 7 | 738 | 85 | ||
| GenBank mRNA | AF426314.1 | - | 6 - 7 | 635 | 85 | |
| AK152409.1 | - | 6 - 7 | 668 | 85 | ||
| AK155248.1 | - | 6 - 7 | 663 | 85 | ||
| AK167762.1 | - | 6 - 7 | 711 | 85 | ||
| BC024477.1 | - | 6 - 7 | 653 | 4977001 | 85 | |
| EF676083.1 | - | 6 - 7 | 714 | 85 | ||
More Information
Set Membership: |
|

Sh3glb2
Genomic Map