Assay Details
| Gene Symbol: Trib3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175093.2 | NP_780302.2 | 3 - 4 | 794 | 63 | |
| GenBank mRNA | AF358868.1 | - | 3 - 4 | 771 | 63 | |
| AJ514260.1 | - | 3 - 4 | 719 | 63 | ||
| AK089931.1 | - | 3 - 4 | 670 | 63 | ||
| AK132257.1 | - | 3 - 4 | 733 | 63 | ||
| AK144752.1 | - | 3 - 4 | 713 | 63 | ||
| AK146340.1 | - | 3 - 4 | 723 | 63 | ||
| AK146421.1 | - | 3 - 4 | 652 | 63 | ||
| AK159760.1 | - | 3 - 4 | 734 | 63 | ||
| AK160099.1 | - | 3 - 4 | 785 | 63 | ||
| BC012955.1 | - | 3 - 4 | 766 | 3980838 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
negative regulation of protein kinase activity
apoptotic process
regulation of glucose transport
positive regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to insulin stimulus
response to endoplasmic reticulum stress
regulation of MAP kinase activity
negative regulation of fat cell differentiation
negative regulation of fatty acid biosynthetic process
positive regulation of protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of ubiquitin-protein transferase activity
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
protein kinase activity
protein kinase inhibitor activity
protein binding
ATP binding
transferase activity, transferring phosphorus-containing groups
enzyme binding
protein kinase binding
mitogen-activated protein kinase kinase binding
ubiquitin protein ligase binding
ubiquitin-protein transferase regulator activity
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
negative regulation of protein kinase activity
apoptotic process
regulation of glucose transport
positive regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to insulin stimulus
response to endoplasmic reticulum stress
regulation of MAP kinase activity
negative regulation of fat cell differentiation
negative regulation of fatty acid biosynthetic process
positive regulation of protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of ubiquitin-protein transferase activity
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
protein kinase activity
protein kinase inhibitor activity
protein binding
ATP binding
transferase activity, transferring phosphorus-containing groups
enzyme binding
protein kinase binding
mitogen-activated protein kinase kinase binding
ubiquitin protein ligase binding
ubiquitin-protein transferase regulator activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21748856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21748853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21748857 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009988 | BC012955.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009988 | AK146340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009988 | AK132257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009988 | NM_175093.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159195 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173096 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009988 | BC012955.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009988 | AK146340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009988 | AK132257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009988 | NM_175093.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331570 | BC012955.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21748852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21748849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21748854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21748846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21748843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21748850 | - | - |
| Whole Mouse Genome G4122A | A_51_P331570 | BC012955.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P600274 | AK160099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P600274 | BC012955.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P600274 | AK146340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P600274 | AK146421.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P600274 | AK159760.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01751515 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02265602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01751516 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01552231 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01751518 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01552233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02265603 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21748856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21748853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21748857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21748794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01159559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01552234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02051408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01253403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01178388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01597062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01178389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02265604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02054622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01762396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01320640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01401957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21748824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02196932 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10488610 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488609 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488611 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488612 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488614 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10488613 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426065_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456225_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488608 | - | - |
| Mouse Genome 430 2.0 Array | 1426065_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456225_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1426065_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456225_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228775_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228775_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200041587.mm.1 | NM_175093.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041587.mm.1 | NM_175093.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041587.mm.1 | TC0200004896.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041588.mm.1 | NM_175093.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041588.mm.1 | NM_175093.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041588.mm.1 | TC0200004896.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041589.mm.1 | NM_175093.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041589.mm.1 | NM_175093.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041589.mm.1 | TC0200004896.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200041590.mm.1 | NM_175093.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041590.mm.1 | NM_175093.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200041590.mm.1 | TC0200004896.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456225_x_at | - | - |
| Mouse Expression Set 430 | 1426065_a_at | - | - |
| Mouse Expression Set 430 | 1426065_PM_a_at | - | - |
| Mouse Expression Set 430 | 1456225_PM_x_at | - | - |
| Murine Genome U74v2 Set | 161067_at | - | - |
| Murine Genome U74v2 Set | 161067_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456225_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426065_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426065_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456225_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456225_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426065_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456225_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426065_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4850670 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5890161 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3710722 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252762 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244279 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225528 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244279 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225528 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252762 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003108.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850670 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003108.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850670 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1252762 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225528 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244279 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252762 | - | - |

Trib3
Genomic Map