Assay Details
| Gene Symbol: Myh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039545.2 | NP_001034634.2 | 22 - 23 | 2821 | 116 | |
| GenBank mRNA | BC008538.1 | - | 21 - 22 | 2760 | 3495412 | 116 |
More Information
Updated Mapping Information
Set Membership: |
|

Myh2
Genomic Map