Assay Details
| Gene Symbol: Mgat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145128.3 | NP_660110.2 | 13 - 14 | 2162 | 76 | |
| GenBank mRNA | AF474154.1 | - | 11 - 12 | 1542 | 76 | |
| AF474155.1 | - | 11 - 12 | 1605 | 76 | ||
| AK030402.1 | - | 13 - 14 | 2130 | 76 | ||
| BC125533.1 | - | 11 - 12 | 1570 | 40130048 | 76 | |
| BC125535.1 | - | 11 - 12 | 1570 | 40130050 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgat5
Genomic Map