Assay Details
| Gene Symbol: Nek9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145138.2 | NP_660120.2 | 10 - 11 | 1329 | 72 | |
| GenBank mRNA | AJ489828.1 | - | 10 - 11 | 1304 | 72 | |
| AK173323.1 | - | 10 - 11 | 1313 | 72 | ||
| BC117971.2 | - | 10 - 11 | 1270 | 40098375 | 72 | |
| BC117972.2 | - | 10 - 11 | 1269 | 40098378 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek9
Genomic Map