Assay Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145400.3 | NP_663375.3 | 2 - 3 | 209 | 81 | |
| GenBank mRNA | AK028721.1 | - | 2 - 3 | 129 | 81 | |
| AK028850.1 | - | 2 - 3 | 167 | 81 | ||
| AK172893.1 | - | 2 - 3 | 169 | 81 | ||
| BC021406.1 | - | 2 - 3 | 158 | 81 | ||
| BC025043.1 | - | 2 - 3 | 121 | 81 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_145400.3 transcripts.
Set Membership: |
|

Ube4a
Genomic Map