Assay Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145400.3 | NP_663375.3 | 4 - 5 | 543 | 105 | |
| GenBank mRNA | AK028721.1 | - | 4 - 5 | 520 | 105 | |
| AK028850.1 | - | 4 - 5 | 558 | 105 | ||
| AK172893.1 | - | 4 - 5 | 503 | 105 | ||
| BC006649.1 | - | 5 - 6 | 682 | 3498834 | 105 | |
| BC021406.1 | - | 4 - 5 | 549 | 105 | ||
| BC025043.1 | - | 4 - 5 | 512 | 105 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC006649.1 transcripts.
Set Membership: |
|

Ube4a
Genomic Map