Assay Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145400.3 | NP_663375.3 | 7 - 8 | 975 | 102 | |
| GenBank mRNA | AK028721.1 | - | 7 - 8 | 952 | 102 | |
| AK028850.1 | - | 7 - 8 | 990 | 102 | ||
| AK172893.1 | - | 7 - 8 | 935 | 102 | ||
| BC006649.1 | - | 8 - 9 | 1114 | 3498834 | 102 | |
| BC021406.1 | - | 7 - 8 | 981 | 102 | ||
| BC025043.1 | - | 7 - 8 | 944 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube4a
Genomic Map