Assay Details
| Gene Symbol: Ubl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145405.2 | NP_663380.1 | 1 - 2 | 130 | 65 | |
| GenBank mRNA | AJ627038.1 | - | 1 - 2 | 91 | 65 | |
| AK146110.1 | - | 120 | 65 | |||
| BC010817.1 | - | 1 - 2 | 109 | 4215699 | 65 | |
| BM932729.1 | - | 1 - 2 | 104 | 65 | ||
| Gene Symbol: Gm38419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278271.1 | NP_001265200.1 | 4 - 5 | 729 | 65 | |
| NM_001278272.1 | NP_001265201.1 | 4 - 5 | 693 | 65 | ||
| NR_103489.1 | 4 - 5 | 729 | 65 | |||
| GenBank mRNA | EU599043.1 | - | 4 - 5 | 598 | 65 | |
| EU599044.1 | - | 4 - 5 | 562 | 65 | ||
| EU599045.1 | - | 4 - 5 | 598 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467745 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467744 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467742 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467743 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P637730 | NM_145405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P637730 | AK048689.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P637730 | AK146110.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P637730 | BC010817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P637730 | NM_145405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P637730 | AK048689.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P637730 | AK146110.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P637730 | BC010817.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898416 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 62755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864761 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65927 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168657 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P637730 | NM_145405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P637730 | AK048689.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P637730 | AK146110.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P637730 | BC010817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P637730 | NM_145405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P637730 | AK048689.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P637730 | AK146110.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P637730 | BC010817.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467736 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168657 | - | - |
| Whole Mouse Genome G4122A | A_52_P637730 | NM_145405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637730 | AK048689.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637730 | AK146110.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637730 | BC010817.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637730 | NM_145405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637730 | AK048689.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637730 | AK146110.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P637730 | BC010817.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147056 | AK048689.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127146 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168657 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14635117 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14980265 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467743 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467745 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467744 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14824176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14824170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14980266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14824156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467741 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10605321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605325 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605327 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424538_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424539_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10605319 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10605324 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10605319 | - | - |
| Mouse Genome 430 2.0 Array | 1424538_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424539_at | - | - |
| Mouse Genome 430 2.0 Array | 1424539_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424538_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27643_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27643_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00015596.mm.1 | NM_145405.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015596.mm.1 | NM_145405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015596.mm.1 | NR_045779.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015596.mm.1 | NR_045779.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015596.mm.1 | TC0X00003434.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00015594.mm.1 | NM_145405.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015594.mm.1 | NM_145405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015594.mm.1 | NR_045779.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015594.mm.1 | NR_045779.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015594.mm.1 | TC0X00003434.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00015603.mm.1 | NM_145405.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015603.mm.1 | NM_145405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015603.mm.1 | NR_045779.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015603.mm.1 | NR_045779.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015603.mm.1 | TC0X00003434.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00015602.mm.1 | NM_145405.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015602.mm.1 | NM_145405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015602.mm.1 | NR_045779.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015602.mm.1 | NR_045779.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015602.mm.1 | TC0X00003434.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00015601.mm.1 | NM_145405.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015601.mm.1 | NM_145405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015601.mm.1 | TC0X00003434.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424539_PM_at | - | - |
| Mouse Expression Set 430 | 1424538_PM_at | - | - |
| Mouse Expression Set 430 | 1424539_at | - | - |
| Mouse Expression Set 430 | 1424538_at | - | - |
| Murine Genome U74v2 Set | 160856_PM_at | - | - |
| Murine Genome U74v2 Set | 104019_PM_at | - | - |
| Murine Genome U74v2 Set | 160856_at | - | - |
| Murine Genome U74v2 Set | 104019_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424538_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424538_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424539_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424539_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424538_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424539_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424538_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424539_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 362464 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4780039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4780039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2466034 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2466034 | - | - |
| mouse-6 v1.1 (Illumina) | scl027643.2_36-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl027643.2_36-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2466034 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2466034 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl027643.2_36-S | - | - |

Ubl4
Genomic Map