Assay Details
| Gene Symbol: Sds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145565.1 | NP_663540.1 | 1 - 2 | 46 | 92 | |
| GenBank mRNA | AK149438.1 | - | 1 - 2 | 63 | 92 | |
| BC011259.1 | - | 1 - 2 | 43 | 92 | ||
| BC021605.1 | - | 1 - 2 | 43 | 5097741 | 92 | |
| BC021950.1 | - | 1 - 2 | 46 | 5102037 | 92 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK149438.1,BC021605.1,BC021950.1,NM_145565.1 transcripts.
Set Membership: |
|

Sds
Genomic Map