Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 10 - 11 | 1438 | 96 | |
| GenBank mRNA | AK031767.1 | - | 10 - 11 | 1417 | 96 | |
| AK049053.1 | - | 10 - 11 | 1852 | 96 | ||
| AK169116.1 | - | 2 - 3 | 261 | 96 | ||
| AK220478.1 | - | 9 - 10 | 1151 | 96 | ||
| BC005470.1 | - | 8 - 9 | 1007 | 3496607 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp11
Genomic Map