Assay Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145628.4 | NP_663603.3 | 15 - 16 | 2146 | 82 | |
| GenBank mRNA | AK031767.1 | - | 15 - 16 | 2125 | 82 | |
| AK169116.1 | - | 7 - 8 | 969 | 82 | ||
| AK172230.1 | - | 2 - 3 | 1109 | 82 | ||
| AK220478.1 | - | 13 - 14 | 2661 | 82 | ||
| BC005470.1 | - | 13 - 14 | 1715 | 3496607 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp11
Genomic Map