Assay Details
| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145711.4 | NP_663757.3 | 4 - 5 | 925 | 62 | |
| GenBank mRNA | AF472514.1 | - | 4 - 5 | 721 | 62 | |
| AK029694.1 | - | 4 - 5 | 722 | 62 | ||
| AK038671.1 | - | 4 - 5 | 928 | 62 | ||
| AK133397.1 | - | 4 - 5 | 722 | 62 | ||
| AK161578.1 | - | 4 - 5 | 710 | 62 | ||
| BC080732.1 | - | 4 - 5 | 842 | 6518356 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tox
Genomic Map