Assay Details
| Gene Symbol: Tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146214.3 | NP_666326.1 | 1 - 2 | 88 | 76 | |
| GenBank mRNA | AK090244.1 | - | 1 - 2 | 86 | 76 | |
| AK149383.1 | - | 1 - 2 | 86 | 76 | ||
| AK149526.1 | - | 1 - 2 | 86 | 76 | ||
| BC023949.1 | - | 1 - 2 | 72 | 5310059 | 76 | |
| BC024120.1 | - | 1 - 2 | 63 | 5097591 | 76 | |
| BC028821.1 | - | 1 - 2 | 63 | 5097589 | 76 | |
| BC030728.1 | - | 1 - 2 | 67 | 5099036 | 76 | |
| BC030729.1 | - | 1 - 2 | 61 | 5099570 | 76 | |
| BC037526.1 | - | 1 - 2 | 43 | 5097795 | 76 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK090244.1,AK149383.1,AK149526.1,BC023949.1,BC024120.1,BC028821.1,BC030728.1,BC030729.1,BC037526.1,NM_146214.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Hyporesponsiveness to glucocorticoids in mice genetically deficient for the corticosteroid binding globulin.
- Authors
- Petersen HH, Andreassen TK, Breiderhoff T, Brasen JH, Schulz H, Gross V, Grone HJ, Nykjaer A, Willnow TE
- Journal
- Mol Cell Biol - Page No 7236 (Sun Oct 01 00:00:00 EDT 2006)
- Volume (Year)
- 26 (2006)
- Product Usage
- -

Tat
Genomic Map