Assay Details
| Gene Symbol: Tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146214.3 | NP_666326.1 | 4 - 5 | 503 | 72 | |
| GenBank mRNA | AK090244.1 | - | 4 - 5 | 501 | 72 | |
| AK149383.1 | - | 4 - 5 | 501 | 72 | ||
| AK149526.1 | - | 4 - 5 | 501 | 72 | ||
| BC023949.1 | - | 4 - 5 | 487 | 5310059 | 72 | |
| BC024120.1 | - | 4 - 5 | 478 | 5097591 | 72 | |
| BC024264.1 | - | 3 - 4 | 426 | 5098682 | 72 | |
| BC025934.1 | - | 4 - 5 | 438 | 5097113 | 72 | |
| BC028821.1 | - | 4 - 5 | 478 | 5097589 | 72 | |
| BC030728.1 | - | 4 - 5 | 482 | 5099036 | 72 | |
| BC030729.1 | - | 4 - 5 | 476 | 5099570 | 72 | |
| BC037526.1 | - | 4 - 5 | 458 | 5097795 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tat
Genomic Map