Assay Details
| Gene Symbol: Tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146214.3 | NP_666326.1 | 6 - 7 | 800 | 80 | |
| GenBank mRNA | AK149383.1 | - | 6 - 7 | 798 | 80 | |
| AK149526.1 | - | 6 - 7 | 798 | 80 | ||
| BC023949.1 | - | 6 - 7 | 784 | 5310059 | 80 | |
| BC024120.1 | - | 6 - 7 | 775 | 5097591 | 80 | |
| BC024264.1 | - | 5 - 6 | 723 | 5098682 | 80 | |
| BC025934.1 | - | 6 - 7 | 735 | 5097113 | 80 | |
| BC028821.1 | - | 6 - 7 | 775 | 5097589 | 80 | |
| BC030728.1 | - | 6 - 7 | 779 | 5099036 | 80 | |
| BC030729.1 | - | 6 - 7 | 773 | 5099570 | 80 | |
| BC037526.1 | - | 6 - 7 | 755 | 5097795 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tat
Genomic Map