Assay Details
| Gene Symbol: Als2cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146059.1 | NP_001139531.1 | 14 - 15 | 1630 | 63 | |
| NM_001146060.1 | NP_001139532.1 | 14 - 15 | 1710 | 63 | ||
| NM_146228.4 | NP_666340.3 | 14 - 15 | 1627 | 63 | ||
| GenBank mRNA | AB107016.1 | - | 14 - 15 | 1627 | 63 | |
| AF481142.1 | - | 13 - 14 | 1523 | 63 | ||
| AK030815.1 | - | 7 - 8 | 764 | 63 | ||
| AK053076.1 | - | 14 - 15 | 1600 | 63 | ||
| AK131149.1 | - | 14 - 15 | 1629 | 63 | ||
| AK155451.1 | - | 14 - 15 | 1696 | 63 | ||
| BC117928.1 | - | 13 - 14 | 1580 | 40097635 | 63 | |
| BC117929.1 | - | 13 - 14 | 1577 | 40097636 | 63 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK030815.1 transcripts.
Set Membership: |
|

Als2cl
Genomic Map