Assay Details
| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146239.2 | NP_666351.2 | 15 - 16 | 1814 | 71 | |
| GenBank mRNA | BC031778.1 | - | 1361 | 4501115 | 71 | |
| BC049904.1 | - | 1361 | 4501115 | 71 | ||
| BC064815.1 | - | 15 - 16 | 1799 | 6516981 | 71 | |
| BQ442862.1 | - | 5 - 6 | 398 | 71 | ||
More Information
Set Membership: |
|

Cdk17
Genomic Map