Assay Details
| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146241.2 | NP_666353.1 | 4 - 5 | 1401 | 72 | |
| GenBank mRNA | AK029833.1 | - | 731 | 72 | ||
| AK034093.1 | - | 4 - 5 | 780 | 72 | ||
| AK163726.1 | - | 2339 | 72 | |||
| BC032288.1 | - | 4 - 5 | 1406 | 5368580 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trhde
Genomic Map