Assay Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147201.2 | NP_671734.1 | 11 - 12 | 1142 | 68 | |
| GenBank mRNA | AF302138.1 | - | 11 - 12 | 1141 | 68 | |
| AK046142.1 | - | 12 - 13 | 1160 | 68 | ||
| AK077440.1 | - | 11 - 12 | 1178 | 68 | ||
| AK170501.1 | - | 11 - 12 | 1158 | 68 | ||
| AK188331.1 | - | 3 - 4 | 283 | 68 | ||
| AK207083.1 | - | 3 - 4 | 283 | 68 | ||
| BC004756.1 | - | 11 - 12 | 1112 | 3154089 | 68 | |
| BC018463.1 | - | 11 - 12 | 1116 | 3598438 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrbp1
Genomic Map