Assay Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147201.2 | NP_671734.1 | 12 - 13 | 1241 | 63 | |
| GenBank mRNA | AF302138.1 | - | 12 - 13 | 1240 | 63 | |
| AK046142.1 | - | 13 - 14 | 1259 | 63 | ||
| AK170501.1 | - | 12 - 13 | 1257 | 63 | ||
| AK182568.1 | - | 1 - 2 | 64 | |||
| AK185308.1 | - | 1 - 2 | 28 | 63 | ||
| AK196002.1 | - | 1 - 2 | 28 | 63 | ||
| AK215481.1 | - | 1 - 2 | 87 | 63 | ||
| BC004756.1 | - | 12 - 13 | 1211 | 3154089 | 63 | |
| BC013212.1 | - | 1 - 2 | 635 | 63 | ||
| BC018463.1 | - | 12 - 13 | 1215 | 3598438 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrbp1
Genomic Map