Assay Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148932.2 | NP_683734.2 | 3 - 4 | 985 | 53 | |
| GenBank mRNA | AF516680.1 | - | 3 - 4 | 949 | 53 | |
| AK154473.1 | - | 3 - 4 | 986 | 53 | ||
| AK172982.1 | - | 2 - 3 | 363 | 53 | ||
| BC053101.1 | - | 3 - 4 | 961 | 6405486 | 53 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24116186 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24116190 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24116189 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959056 | NM_148932.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959056 | AF516680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959056 | AK154473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959056 | BC028835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959056 | CF545814.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562256 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149526 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119167 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153525 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959056 | NM_148932.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959056 | AF516680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959056 | AK154473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959056 | BC028835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959056 | CF545814.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116181 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153525 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05755479 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119167 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05755455 | - | - |
| Whole Mouse Genome G4122A | A_51_P191393 | NM_148932.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P191393 | AF516680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P191393 | AK154473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P191393 | BC053101.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P191393 | AK172982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407751 | NM_148932.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407751 | AF516680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407751 | AK154473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407751 | BC028835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P407751 | W44309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P576280 | NM_148932.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P576280 | AF516680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P576280 | AK154473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P576280 | BC053101.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P576280 | AK172982.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102871 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119167 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153525 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04601916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24116186 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24116189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24116190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04881848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04516955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04471379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04908385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04351461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04698061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04765593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04296910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04782005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04851575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04725259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04725258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04471380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04391228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04588279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116195 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534442 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534448 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420004_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427886_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534441 | - | - |
| Mouse Genome 430 2.0 Array | 1427886_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420004_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1420004_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1427886_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107939_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107939_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500030671.mm.1 | TC0500003393.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030671.mm.1 | NM_148932.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030671.mm.1 | NM_148932.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030670.mm.1 | TC0500003393.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030670.mm.1 | NM_148932.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030670.mm.1 | NM_148932.2 | Refseq |
| Mouse Expression Set 430 | 1427886_at | - | - |
| Mouse Expression Set 430 | 1427886_PM_at | - | - |
| Murine Genome U74v2 Set | 96174_PM_at | - | - |
| Murine Genome U74v2 Set | 96174_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427886_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427886_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420004_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427886_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427886_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420004_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 709146 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 70152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 70152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232596 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232596 | - | - |
| mouse-6 v1.1 (Illumina) | scl25927.13_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70152 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25927.13_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232596 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232596 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25927.13_1-S | - | - |

Pom121
Genomic Map