Assay Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148932.2 | NP_683734.2 | 4 - 5 | 1073 | 86 | |
| GenBank mRNA | AF516680.1 | - | 4 - 5 | 1037 | 86 | |
| AK154473.1 | - | 4 - 5 | 1074 | 86 | ||
| AK172982.1 | - | 3 - 4 | 451 | 86 | ||
| BC053101.1 | - | 4 - 5 | 1049 | 6405486 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pom121
Genomic Map