Assay Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148932.2 | NP_683734.2 | 10 - 11 | 1805 | 92 | |
| GenBank mRNA | AF516680.1 | - | 10 - 11 | 1766 | 92 | |
| AK154473.1 | - | 10 - 11 | 1806 | 92 | ||
| AK172982.1 | - | 9 - 10 | 1183 | 92 | ||
| BC053101.1 | - | 10 - 11 | 1781 | 6405486 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pom121
Genomic Map