Assay Details
| Gene Symbol: Osbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152818.2 | NP_690031.2 | 11 - 12 | 2218 | 66 | |
| GenBank mRNA | AK007088.1 | - | 7 - 8 | 799 | 66 | |
| AK044598.1 | - | 8 - 9 | 1007 | 66 | ||
| BC031794.1 | - | 9 - 10 | 1181 | 4501119 | 66 | |
| BC034567.1 | - | 1 - 2 | 58 | 66 | ||
| BC051185.1 | - | 9 - 10 | 1178 | 66 | ||
| BC058356.1 | - | 10 - 11 | 1361 | 5696357 | 66 | |
| BC058602.1 | - | 9 - 10 | 1181 | 6467653 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbp2
Genomic Map