Assay Details
| Gene Symbol: Bgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007542.4 | NP_031568.2 | 4 - 5 | 708 | 67 | |
| GenBank mRNA | AK135496.1 | - | 4 - 5 | 709 | 67 | |
| AK155973.1 | - | 4 - 5 | 709 | 67 | ||
| AK164867.1 | - | 4 - 5 | 709 | 67 | ||
| AK171870.1 | - | 4 - 5 | 709 | 67 | ||
| BC005452.1 | - | 4 - 5 | 702 | 3594469 | 67 | |
| BC019502.1 | - | 4 - 5 | 711 | 4237268 | 67 | |
| BC052857.1 | - | 4 - 5 | 693 | 30061850 | 67 | |
| L20276.1 | - | 4 - 5 | 708 | 67 | ||
| X53928.1 | - | 803 | 67 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Bgn
Genomic Map