Assay Details
| Gene Symbol: Bgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007542.4 | NP_031568.2 | 6 - 7 | 912 | 88 | |
| GenBank mRNA | AK135496.1 | - | 6 - 7 | 913 | 88 | |
| AK164867.1 | - | 6 - 7 | 913 | 88 | ||
| AK171870.1 | - | 6 - 7 | 913 | 88 | ||
| BC005452.1 | - | 6 - 7 | 906 | 3594469 | 88 | |
| BC019502.1 | - | 6 - 7 | 915 | 4237268 | 88 | |
| BC052857.1 | - | 6 - 7 | 897 | 30061850 | 88 | |
| L20276.1 | - | 6 - 7 | 912 | 88 | ||
| X53928.1 | - | 1007 | 88 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Bgn
Genomic Map