Assay Details
| Gene Symbol: Bscl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136064.3 | NP_001129536.1 | 6 - 7 | 1285 | 88 | |
| NM_001290823.1 | NP_001277752.1 | 5 - 6 | 744 | 88 | ||
| NM_008144.5 | NP_032170.3 | 5 - 6 | 1135 | 88 | ||
| GenBank mRNA | AB030196.1 | - | 5 - 6 | 754 | 88 | |
| AF069954.2 | - | 5 - 6 | 960 | 88 | ||
| AK004634.1 | - | 3 - 4 | 2834 | 88 | ||
| AK151606.1 | - | 5 - 6 | 713 | 88 | ||
| AK155753.1 | - | 4 - 5 | 1252 | 88 | ||
| AK158901.1 | - | 5 - 6 | 1136 | 88 | ||
| AK170183.1 | - | 5 - 6 | 711 | 88 | ||
| BC043023.1 | - | 5 - 6 | 899 | 5689013 | 88 | |
| BC060048.1 | - | 4 - 5 | 1965 | 88 | ||
| BC061689.1 | - | 6 - 7 | 1023 | 6306706 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bscl2
Genomic Map