Assay Details
| Gene Symbol: Hdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008230.6 | NP_032256.3 | 9 - 10 | 1166 | 71 | |
| GenBank mRNA | AF109137.1 | - | 9 - 10 | 1076 | 71 | |
| AK088545.1 | - | 9 - 10 | 1153 | 71 | ||
| AK133455.1 | - | 9 - 10 | 1167 | 71 | ||
| AK150168.1 | - | 9 - 10 | 1154 | 71 | ||
| AK153104.1 | - | 9 - 10 | 1154 | 71 | ||
| X57437.1 | - | 9 - 10 | 1131 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdc
Genomic Map