Assay Details
| Gene Symbol: Lgals7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008496.4 | NP_032522.2 | 3 - 4 | 393 | 68 | |
| GenBank mRNA | AF038562.1 | - | 318 | 68 | ||
| AF331640.1 | - | 2 - 3 | 298 | 68 | ||
| AK003153.1 | - | 3 - 4 | 327 | 68 | ||
| AK008773.1 | - | 3 - 4 | 327 | 68 | ||
| AK009151.1 | - | 3 - 4 | 336 | 68 | ||
| BC116838.1 | - | 3 - 4 | 354 | 40126157 | 68 | |
| BC116840.1 | - | 3 - 4 | 354 | 40126159 | 68 | |
More Information
Set Membership: |
|

Lgals7
Genomic Map