Assay Details
| Gene Symbol: Blnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008528.4 | NP_032554.2 | 7 - 8 | 1083 | 108 | |
| GenBank mRNA | AB015290.1 | - | 7 - 8 | 757 | 108 | |
| AF068182.1 | - | 7 - 8 | 738 | 108 | ||
| AJ222814.1 | - | 7 - 8 | 110 | |||
| AK089704.1 | - | 7 - 8 | 724 | 109 | ||
| BC059785.1 | - | 7 - 8 | 709 | 5339723 | 108 | |
| Y17159.1 | - | 7 - 8 | 1083 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Blnk
Genomic Map