Assay Details
| Gene Symbol: Blnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008528.4 | NP_032554.2 | 9 - 10 | 1218 | 112 | |
| GenBank mRNA | AB015290.1 | - | 9 - 10 | 892 | 112 | |
| AF068182.1 | - | 9 - 10 | 873 | 112 | ||
| AK089704.1 | - | 9 - 10 | 859 | 112 | ||
| BC059785.1 | - | 9 - 10 | 844 | 5339723 | 112 | |
| Y17159.1 | - | 9 - 10 | 1218 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Blnk
Genomic Map