Assay Details
| Gene Symbol: Orc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025378.2 | NP_001020549.1 | 10 - 11 | 1011 | 106 | |
| NM_001271526.1 | NP_001258455.1 | 13 - 14 | 1536 | 106 | ||
| NM_008765.3 | NP_032791.1 | 13 - 14 | 1542 | 106 | ||
| GenBank mRNA | AB190256.1 | - | 10 - 11 | 1001 | 106 | |
| AK048408.1 | - | 13 - 14 | 1361 | 106 | ||
| AK051747.1 | - | 13 - 14 | 1371 | 106 | ||
| AK133604.1 | - | 13 - 14 | 1362 | 106 | ||
| AK146281.1 | - | 13 - 14 | 1361 | 106 | ||
| AK146859.1 | - | 13 - 14 | 1356 | 106 | ||
| BC015257.1 | - | 13 - 14 | 1352 | 4234871 | 106 | |
| U27457.1 | - | 13 - 14 | 1361 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Orc2
Genomic Map