Assay Details
| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282017.1 | NP_001268946.1 | 1 - 2 | 108 | 62 | |
| NM_008951.2 | NP_032977.1 | 1 - 2 | 108 | 62 | ||
| GenBank mRNA | AB029142.1 | - | 1 - 2 | 39 | 62 | |
| AB029143.1 | - | 1 - 2 | 39 | 62 | ||
| AB029144.1 | - | 1 - 2 | 37 | 62 | ||
| AB029145.1 | - | 1 - 2 | 39 | 62 | ||
| AF013099.1 | - | 1 - 2 | 39 | 62 | ||
| AF175574.1 | - | 1 - 2 | 39 | 62 | ||
| AK145524.1 | - | 1 - 2 | 47 | 62 | ||
| BC009005.1 | - | 1 - 2 | 57 | 3581937 | 62 | |
| BC016433.1 | - | 1 - 2 | 42 | 62 | ||
| BY099585.1 | - | 1 - 2 | 109 | 62 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB029142.1,AB029143.1,AB029144.1,AB029145.1,AF013099.1,AF175574.1,AK145524.1,BC009005.1,NM_001282017.1,NM_008951.2 transcripts.
Set Membership: |
|

Psmd4
Genomic Map