Assay Details
| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008951.2 | NP_032977.1 | 7 - 8 | 842 | 62 | |
| GenBank mRNA | AB029142.1 | - | 7 - 8 | 773 | 62 | |
| AB029145.1 | - | 7 - 8 | 773 | 62 | ||
| AF013099.1 | - | 7 - 8 | 773 | 62 | ||
| AK010969.1 | - | 7 - 8 | 771 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmd4
Genomic Map