Assay Details
| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282017.1 | NP_001268946.1 | 8 - 9 | 987 | 64 | |
| NM_008951.2 | NP_032977.1 | 8 - 9 | 978 | 64 | ||
| GenBank mRNA | AB029142.1 | - | 8 - 9 | 909 | 64 | |
| AB029143.1 | - | 8 - 9 | 918 | 64 | ||
| AB029144.1 | - | 8 - 9 | 916 | 64 | ||
| AF013099.1 | - | 8 - 9 | 909 | 64 | ||
| AF175574.1 | - | 8 - 9 | 918 | 64 | ||
| AK010969.1 | - | 8 - 9 | 907 | 64 | ||
| AK145524.1 | - | 8 - 9 | 926 | 64 | ||
| BC009005.1 | - | 8 - 9 | 936 | 3581937 | 64 | |
| BC016433.1 | - | 5 - 6 | 1777 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmd4
Genomic Map