Assay Details
| Gene Symbol: Psmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009439.1 | NP_033465.1 | 4 - 5 | 845 | 88 | |
| GenBank mRNA | AK031366.1 | - | 4 - 5 | 835 | 88 | |
| AK077252.1 | - | 4 - 5 | 794 | 88 | ||
| AK089933.1 | - | 4 - 5 | 803 | 88 | ||
| AK145467.1 | - | 4 - 5 | 835 | 88 | ||
| AK145488.1 | - | 4 - 5 | 834 | 88 | ||
| AK153505.1 | - | 4 - 5 | 762 | 88 | ||
| AK164333.1 | - | 4 - 5 | 809 | 88 | ||
| AK164593.1 | - | 4 - 5 | 834 | 88 | ||
| AK165520.1 | - | 4 - 5 | 780 | 88 | ||
| AK166683.1 | - | 4 - 5 | 805 | 88 | ||
| AK166714.1 | - | 4 - 5 | 833 | 88 | ||
| AK167005.1 | - | 4 - 5 | 820 | 88 | ||
| AK167285.1 | - | 4 - 5 | 800 | 88 | ||
| AK169690.1 | - | 4 - 5 | 834 | 88 | ||
| BC003197.1 | - | 4 - 5 | 845 | 3586427 | 88 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK003024.1 | - | Chr. - - [-] on Build | 88 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Psmd3
Genomic Map